Assay Details
| Gene Symbol: POM121L12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002187.1 | XP_009000435.1 | 2 - 3 | 237 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POM121L12| Gene Symbol: POM121L12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002187.1 | XP_009000435.1 | 2 - 3 | 237 | 77 | |
Set Membership: |
|