Assay Details
| Gene Symbol: C11H11orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978443.1 | XP_017833932.1 | 3 - 4 | 1035 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11H11orf45| Gene Symbol: C11H11orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978443.1 | XP_017833932.1 | 3 - 4 | 1035 | 92 | |
Set Membership: |
|