Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743116.4 | XP_002743162.1 | 3 - 4 | 2670 | 107 | |
| XM_008999924.2 | XP_008998172.1 | 3 - 4 | 2670 | 107 | ||
| XM_017974936.1 | XP_017830425.1 | 2 - 3 | 2495 | 107 | ||
| XM_017974940.1 | XP_017830429.1 | 2 - 3 | 2495 | 107 | ||
More Information
Set Membership: |
|

TRMO
Genomic Map