Assay Details
| Gene Symbol: ADGRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985949.2 | XP_008984197.1 | 8 - 9 | 3442 | 67 | |
| XM_008985952.2 | XP_008984200.1 | 8 - 9 | 3442 | 67 | ||
| XM_008985953.2 | XP_008984201.1 | 7 - 8 | 773 | 67 | ||
| XM_008985954.2 | XP_008984202.1 | 6 - 7 | 874 | 67 | ||
More Information
Set Membership: |
|

ADGRG3
Genomic Map