Assay Details
| Gene Symbol: SCIMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997266.2 | XP_008995514.1 | 3 - 4 | 294 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCIMP| Gene Symbol: SCIMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997266.2 | XP_008995514.1 | 3 - 4 | 294 | 79 | |