Assay Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998208.2 | XP_008996456.1 | 4 - 5 | 2440 | 107 | |
| XM_017973082.1 | XP_017828571.1 | 3 - 4 | 2323 | 107 | ||
| XM_017973083.1 | XP_017828572.1 | 4 - 5 | 824 | 107 | ||
More Information
Set Membership: |
|

CCDC122
Genomic Map