Assay Details
| Gene Symbol: LOC100409113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971648.1 | XP_017827137.1 | 36 - 37 | 5108 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100409113| Gene Symbol: LOC100409113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971648.1 | XP_017827137.1 | 36 - 37 | 5108 | 106 | |
Set Membership: |
|