Assay Details
| Gene Symbol: GIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978475.1 | XP_017833964.1 | 5 - 6 | 881 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

GIF| Gene Symbol: GIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978475.1 | XP_017833964.1 | 5 - 6 | 881 | 73 | |