Assay Details
| Gene Symbol: ALDOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743150.3 | XP_002743196.1 | 5 - 6 | 681 | 78 | |
| XM_017975082.1 | XP_017830571.1 | 5 - 6 | 681 | 78 | ||
More Information
Set Membership: |
|

ALDOB
Genomic Map