Assay Details
| Gene Symbol: GJD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733306.3 | XP_003733354.1 | 1 - 2 | 2066 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GJD4| Gene Symbol: GJD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733306.3 | XP_003733354.1 | 1 - 2 | 2066 | 102 | |
Set Membership: |
|