Assay Details
| Gene Symbol: NR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978768.1 | XP_017834257.1 | 7 - 8 | 1699 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR5A1| Gene Symbol: NR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978768.1 | XP_017834257.1 | 7 - 8 | 1699 | 63 | |
Set Membership: |
|