Assay Details
| Gene Symbol: MMP23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750187.3 | XP_002750233.1 | 5 - 6 | 1334 | 86 | |
| XM_009000252.2 | XP_008998500.1 | 5 - 6 | 1336 | 86 | ||
More Information
Set Membership: |
|

MMP23B
Genomic Map