Assay Details
| Gene Symbol: KRT75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752509.2 | XP_002752555.1 | 2 - 3 | 927 | 80 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT75| Gene Symbol: KRT75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752509.2 | XP_002752555.1 | 2 - 3 | 927 | 80 | |