Assay Details
| Gene Symbol: SAP30L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988582.2 | XP_008986830.1 | 1 - 2 | 843 | 93 | |
| XM_008988585.2 | XP_008986833.1 | 1 - 2 | 843 | 93 | ||
More Information
Set Membership: |
|

SAP30L
Genomic Map