Assay Details
| Gene Symbol: LOC100414476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992123.1 | XP_008990371.1 | 4 - 5 | 358 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100414476| Gene Symbol: LOC100414476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992123.1 | XP_008990371.1 | 4 - 5 | 358 | 121 | |
Set Membership: |
|