Assay Details
| Gene Symbol: PPP1R3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746252.3 | XP_002746298.1 | 1 | 1116 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R3G| Gene Symbol: PPP1R3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746252.3 | XP_002746298.1 | 1 | 1116 | 80 | |
Set Membership: |
|