Assay Details
| Gene Symbol: BEND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735694.1 | XP_003735742.1 | 9 - 10 | 1286 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BEND2| Gene Symbol: BEND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735694.1 | XP_003735742.1 | 9 - 10 | 1286 | 100 | |