Assay Details
| Gene Symbol: TSSK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991680.2 | XP_008989928.1 | 1 | 4420 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSSK1B| Gene Symbol: TSSK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991680.2 | XP_008989928.1 | 1 | 4420 | 77 | |
Set Membership: |
|