Assay Details
| Gene Symbol: APCDD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971669.1 | XP_017827158.1 | 4 - 5 | 1693 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APCDD1L| Gene Symbol: APCDD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971669.1 | XP_017827158.1 | 4 - 5 | 1693 | 84 | |
Set Membership: |
|