Assay Details
| Gene Symbol: SCML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747031.3 | XP_002747077.1 | 7 - 8 | 1370 | 56 | |
| XM_008994862.2 | XP_008993110.1 | 3 - 4 | 1353 | 56 | ||
More Information
Set Membership: |
|

SCML4
Genomic Map