Assay Details
| Gene Symbol: LOC100390922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757393.4 | XP_002757439.3 | 2 - 3 | 709 | 96 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100390922| Gene Symbol: LOC100390922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757393.4 | XP_002757439.3 | 2 - 3 | 709 | 96 | |