Assay Details
| Gene Symbol: LOC100390922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757393.4 | XP_002757439.3 | 7 - 8 | 1192 | 67 | |
| XM_017964075.1 | XP_017819564.1 | 3 - 4 | 130 | 67 | ||
More Information
Set Membership: |
|

LOC100390922
Genomic Map