Assay Details
| Gene Symbol: LOC100390922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757393.4 | XP_002757439.3 | 9 - 10 | 1601 | 63 | |
| XM_017964075.1 | XP_017819564.1 | 5 - 6 | 539 | 63 | ||
More Information
Set Membership: |
|

LOC100390922
Genomic Map