Assay Details
| Gene Symbol: LOC100399608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980266.1 | XP_008978514.1 | 3 - 4 | 308 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100399608| Gene Symbol: LOC100399608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980266.1 | XP_008978514.1 | 3 - 4 | 308 | 79 | |
Set Membership: |
|