Assay Details
| Gene Symbol: PDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984817.2 | XP_008983065.1 | 4 - 5 | 486 | 95 | |
| XM_008984818.2 | XP_008983066.1 | 3 - 4 | 428 | 95 | ||
| XM_008984819.2 | XP_008983067.1 | 3 - 4 | 355 | 95 | ||
| XM_008984820.2 | XP_008983068.1 | 3 - 4 | 402 | 95 | ||
More Information
Set Membership: |
|

PDC
Genomic Map