Assay Details
| Gene Symbol: AMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986946.2 | XP_008985194.1 | 2 - 3 | 1409 | 62 | |
| XM_008986947.2 | XP_008985195.1 | 3 - 4 | 1259 | 62 | ||
More Information
Set Membership: |
|

AMH
Genomic Map