Assay Details
| Gene Symbol: LOC100385873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994754.2 | XP_008993002.2 | 2 - 3 | 152 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100385873| Gene Symbol: LOC100385873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994754.2 | XP_008993002.2 | 2 - 3 | 152 | 89 | |
Set Membership: |
|