Assay Details
| Gene Symbol: GSTT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007964.2 | XP_009006212.1 | 2 - 3 | 266 | 71 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSTT2B| Gene Symbol: GSTT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007964.2 | XP_009006212.1 | 2 - 3 | 266 | 71 | |