Assay Details
| Gene Symbol: CST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747514.4 | XP_002747560.1 | 1 - 2 | 328 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST3| Gene Symbol: CST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747514.4 | XP_002747560.1 | 1 - 2 | 328 | 92 | |