Assay Details
| Gene Symbol: DMRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742915.3 | XP_002742961.1 | 1 - 2 | 566 | 75 | |
| XM_008996345.2 | XP_008994593.1 | 1 - 2 | 566 | 75 | ||
More Information
Set Membership: |
|

DMRT1
Genomic Map