Assay Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744726.3 | XP_002744772.1 | 6 - 7 | 1405 | 57 | |
| XM_008991726.2 | XP_008989974.1 | 6 - 7 | 1428 | 57 | ||
| XM_008991727.2 | XP_008989975.1 | 6 - 7 | 1361 | 57 | ||
More Information
Set Membership: |
|

NUDT12
Genomic Map