Assay Details
| Gene Symbol: MAML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754687.3 | XP_002754733.1 | 2 - 3 | 3461 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAML2| Gene Symbol: MAML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754687.3 | XP_002754733.1 | 2 - 3 | 3461 | 72 | |
Set Membership: |
|