Assay Details
| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981216.2 | XP_008979464.1 | 4 - 5 | 505 | 88 | |
| XM_008981217.2 | XP_008979465.1 | 4 - 5 | 505 | 88 | ||
More Information
Set Membership: |
|

ALKAL2
Genomic Map