Assay Details
| Gene Symbol: LRRC74A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977478.1 | XP_017832967.1 | 4 - 5 | 499 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC74A| Gene Symbol: LRRC74A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977478.1 | XP_017832967.1 | 4 - 5 | 499 | 73 | |