Assay Details
| Gene Symbol: BTN2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746121.3 | XP_002746167.1 | 2 - 3 | 553 | 89 | |
| XM_008993836.2 | XP_008992084.1 | 2 - 3 | 519 | 89 | ||
| XM_008993837.2 | XP_008992085.1 | 2 - 3 | 553 | 89 | ||
| XM_008993840.2 | XP_008992088.1 | 2 - 3 | 553 | 89 | ||
| XM_017971581.1 | XP_017827070.1 | 2 - 3 | 553 | 89 | ||
| XM_017971582.1 | XP_017827071.1 | 2 - 3 | 553 | 89 | ||
More Information
Set Membership: |
|

BTN2A2
Genomic Map