Assay Details
| Gene Symbol: LOC100411501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746372.4 | XP_002746418.3 | 27 - 28 | 3580 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100411501| Gene Symbol: LOC100411501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746372.4 | XP_002746418.3 | 27 - 28 | 3580 | 84 | |
Set Membership: |
|