Assay Details
| Gene Symbol: PNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753719.3 | XP_002753765.2 | 4 - 5 | 629 | 93 | |
| XM_009005660.2 | XP_009003908.1 | 4 - 5 | 597 | 93 | ||
More Information
Set Membership: |
|

PNP
Genomic Map