Assay Details
| Gene Symbol: PROK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758554.3 | XP_002758600.1 | 1 - 2 | 244 | 119 | |
| XM_008982027.2 | XP_008980275.1 | 1 - 2 | 244 | 119 | ||
More Information
Set Membership: |
|

PROK2
Genomic Map