Assay Details
Gene Symbol: AQP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760039.2 | XP_002760085.1 | 2 - 3 | 293 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
