Assay Details
| Gene Symbol: UPK3BL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982013.2 | XP_008980261.1 | 1 - 2 | 180 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPK3BL2| Gene Symbol: UPK3BL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982013.2 | XP_008980261.1 | 1 - 2 | 180 | 89 | |