Assay Details
| Gene Symbol: LOC100393990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967260.1 | XP_017822749.1 | 17 - 18 | 2063 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100393990| Gene Symbol: LOC100393990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967260.1 | XP_017822749.1 | 17 - 18 | 2063 | 90 | |