Assay Details
| Gene Symbol: MGAT4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970056.1 | XP_017825545.1 | 3 - 4 | 312 | 200 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGAT4D| Gene Symbol: MGAT4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970056.1 | XP_017825545.1 | 3 - 4 | 312 | 200 | |
Set Membership: |
|