Assay Details
| Gene Symbol: PRSS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978654.1 | XP_017834143.1 | 2 - 3 | 930 | 78 | |
| XM_017978655.1 | XP_017834144.1 | 2 - 3 | 887 | 78 | ||
| XM_017978656.1 | XP_017834145.1 | 2 - 3 | 288 | 78 | ||
More Information
Set Membership: |
|

PRSS33
Genomic Map