Assay Details
| Gene Symbol: DPEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761084.4 | XP_002761130.2 | 7 - 8 | 1037 | 91 | |
| XM_008986096.2 | XP_008984344.1 | 7 - 8 | 1185 | 91 | ||
More Information
Set Membership: |
|

DPEP2
Genomic Map