Assay Details
| Gene Symbol: NUP50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980055.2 | XP_008978303.1 | 5 - 6 | 1310 | 64 | |
| XM_017964084.1 | XP_017819573.1 | 7 - 8 | 1610 | 64 | ||
| XM_017964089.1 | XP_017819578.1 | 6 - 7 | 1465 | 64 | ||
| XM_017964090.1 | XP_017819579.1 | 5 - 6 | 1310 | 64 | ||
More Information
Set Membership: |
|

NUP50
Genomic Map