Assay Details
| Gene Symbol: C3H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992538.2 | XP_008990786.1 | 1 - 2 | 1999 | 85 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3H4orf45| Gene Symbol: C3H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992538.2 | XP_008990786.1 | 1 - 2 | 1999 | 85 | |