Assay Details
| Gene Symbol: PLA2G2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000549.2 | XP_008998797.1 | 2 - 3 | 257 | 62 | |
| XM_017974379.1 | XP_017829868.1 | 2 - 3 | 266 | 62 | ||
More Information
Set Membership: |
|

PLA2G2D
Genomic Map