Assay Details
| Gene Symbol: C15H3orf30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965227.1 | XP_017820716.1 | 1 - 2 | 3627 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H3orf30| Gene Symbol: C15H3orf30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965227.1 | XP_017820716.1 | 1 - 2 | 3627 | 89 | |