Assay Details
| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966629.1 | XP_017822118.1 | 3 - 4 | 436 | 96 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CR1| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966629.1 | XP_017822118.1 | 3 - 4 | 436 | 96 | |