Assay Details
| Gene Symbol: LOC100395124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752496.3 | XP_002752542.2 | 2 - 3 | 656 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100395124| Gene Symbol: LOC100395124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752496.3 | XP_002752542.2 | 2 - 3 | 656 | 57 | |
Set Membership: |
|